Quintix® Pro Balances
Supplier:   Sartorius
Quintix® Pro Balances
Supplier:  Sartorius
Certificates
Quintix® Pro Balances
36 Options Available Below
Product Details & Documents
Quintix® Pro laboratory balances are not just about meeting expectations but exceeding them. The balances have an array of premium features that enhance usability, flexibility, and incorporate an eco-friendly design.
  • Enhance the user experience with new premium features
  • Offering a wide range of use in different environments and applications
  • Energy efficiency, reduced carbon footprint, and sustainability, minimizing environmental impact from the manufacturing plant to the bench
This laboratory balance is a perfect fit for laboratories seeking a versatile balance that can handle a wide range of tasks. It's an ideal choice for those prioritizing sustainability and reliability in their operations. Moreover, it's a cost-effective solution for budget-conscious laboratories, such as startups or those operating with limited funds. It serves as a great alternative to the Cubis® II laboratory balance.
Specifications for Product Family
Spécifications
  • Catalog No:
  • CA77507-080
  • CA77507-082
  • CA77507-092
  • CA77507-094
  • CA77507-108
  • CA77507-110
  • CA77507-072
  • CA77507-074
  • CA77507-084
  • CA77507-086
  • CA77507-104
  • CA77507-106
  • CA77507-116
  • CA77507-118
  • CA77507-128
  • CA77507-130
  • CA77507-064
  • CA77507-066
  • CA77507-068
  • CA77507-070
  • CA77507-076
  • CA77507-078
  • CA77507-088
  • CA77507-090
  • CA77507-096
  • CA77507-112
  • CA77507-114
  • CA77507-120
  • CA77507-122
  • CA77507-124
  • CA77507-126
  • CA77507-132
  • CA77507-134
  • CA77507-098
  • CA77507-100
  • CA77507-102
  • Operating temperature range:
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • 10…30 °C
  • Plug type:
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Interchangeable country-specific plug-in AC adaptors
  • Communication interface:
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • USB UTL;USB Device
  • Voltage:
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
  • 100 – 240 V
Spécifications
Product Family Options
Analytical models with manual glass draft shield
Informations sur le produit
  • Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • QTX124IMU1S_C
    120 g
    0.06 mg
    Ø 90 mm
    0.1 mg
    ≤1.5 s
    0.08 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,678.37
    $7,678.37
    Each
       $7,678.37
    Supplier Number: QTX124IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-080
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX124IRU1S_C
    120 g
    0.06 mg
    Ø 90 mm
    0.1 mg
    ≤1.5 s
    0.08 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,562.33
    $6,562.33
    Each
       $6,562.33
    Supplier Number: QTX124IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-082
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX224IMU1S_C
    220 g
    0.06 mg
    Ø 90 mm
    0.1 mg
    ≤1.5 s
    0.08 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,649.34
    $8,649.34
    Each
       $8,649.34
    Supplier Number: QTX224IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-092
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX224IRU1S_C
    220 g
    0.06 mg
    Ø 90 mm
    0.1 mg
    ≤1.5 s
    0.08 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,533.29
    $7,533.29
    Each
       $7,533.29
    Supplier Number: QTX224IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-094
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX324IMU1S_C
    320 g
    0.06 mg
    Ø 90 mm
    0.1 mg
    ≤1.5 s
    0.08 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,765.38
    $9,765.38
    Each
       $9,765.38
    Supplier Number: QTX324IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-108
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX324IRU1S_C
    320 g
    0.06 mg
    Ø 90 mm
    0.1 mg
    ≤1.5 s
    0.08 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,649.34
    $8,649.34
    Each
       $8,649.34
    Supplier Number: QTX324IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-110
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
Precision models with manual glass draft shield
Informations sur le produit
  • Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • QTX1203IMU1S_C
    1200 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,794.42
    $8,794.42
    Each
       $8,794.42
    Supplier Number: QTX1203IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-072
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX1203IRU1S_C
    1200 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,678.37
    $7,678.37
    Each
       $7,678.37
    Supplier Number: QTX1203IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-074
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX1503IMU1S_C
    1500 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $9,207.36
    $9,207.36
    Each
       $9,207.36
    Supplier Number: QTX1503IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-084
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX1503IRU1S_C
    1500 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $8,091.32
    $8,091.32
    Each
       $8,091.32
    Supplier Number: QTX1503IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-086
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX323IMU1S_C
    320 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,004.31
    $6,004.31
    Each
       $6,004.31
    Supplier Number: QTX323IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-104
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX323IRU1S_C
    320 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,888.27
    $4,888.27
    Each
       $4,888.27
    Supplier Number: QTX323IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-106
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX423IMU1S_C
    420 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,696.26
    $6,696.26
    Each
       $6,696.26
    Supplier Number: QTX423IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-116
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX423IRU1S_C
    420 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,580.21
    $5,580.21
    Each
       $5,580.21
    Supplier Number: QTX423IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-118
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX623IMU1S_C
    620 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,399.36
    $7,399.36
    Each
       $7,399.36
    Supplier Number: QTX623IMU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-128
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX623IRU1S_C
    620 g
    0.6 mg
    Ø 120 mm
    1 mg
    ≤1.0 s
    0.5 mg
    220×377×346 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,283.31
    $6,283.31
    Each
       $6,283.31
    Supplier Number: QTX623IRU1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-130
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
Precision models without draft shield
Informations sur le produit
  • Modèle
    Weighing Capacity
    Linearity
    Plateau de pesée/taille de la plateforme
    Readability
    Stabilization Time
    Repeatabilty
    Dimensions
    Availability
    Price
  • QTX10201IMO1_C
    10200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,283.31
    $6,283.31
    Each
       $6,283.31
    Supplier Number: QTX10201IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-064
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX10201IRO1_C
    10200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,167.27
    $5,167.27
    Each
       $5,167.27
    Supplier Number: QTX10201IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-066
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX1202IMO1S_C
    1200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,888.27
    $4,888.27
    Each
       $4,888.27
    Supplier Number: QTX1202IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-068
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX1202IRO1S_C
    1200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,772.22
    $3,772.22
    Each
       $3,772.22
    Supplier Number: QTX1202IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-070
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX12201IMO1_C
    12200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,399.36
    $7,399.36
    Each
       $7,399.36
    Supplier Number: QTX12201IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-076
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX12201IRO1_C
    12200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,283.31
    $6,283.31
    Each
       $6,283.31
    Supplier Number: QTX12201IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-078
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX2202IMO1S_C
    2200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,301.21
    $5,301.21
    Each
       $5,301.21
    Supplier Number: QTX2202IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-088
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX2202IRO1S_C
    2200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,185.17
    $4,185.17
    Each
       $4,185.17
    Supplier Number: QTX2202IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-090
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX3201IMO1S_C
    3200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,888.27
    $4,888.27
    Each
       $4,888.27
    Supplier Number: QTX3201IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-096
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX4202IMO1S_C
    4200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,696.26
    $6,696.26
    Each
       $6,696.26
    Supplier Number: QTX4202IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-112
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX4202IRO1S_C
    4200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,580.21
    $5,580.21
    Each
       $5,580.21
    Supplier Number: QTX4202IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-114
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX6201IMO1S_C
    6200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $5,301.21
    $5,301.21
    Each
       $5,301.21
    Supplier Number: QTX6201IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-120
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX6201IRO1S_C
    6200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,185.17
    $4,185.17
    Each
       $4,185.17
    Supplier Number: QTX6201IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-122
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX6202IMO1S_C
    6200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $7,399.36
    $7,399.36
    Each
       $7,399.36
    Supplier Number: QTX6202IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-124
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX6202IRO1S_C
    6200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,283.31
    $6,283.31
    Each
       $6,283.31
    Supplier Number: QTX6202IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-126
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX8201IMO1S_C
    8200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,004.31
    $6,004.31
    Each
       $6,004.31
    Supplier Number: QTX8201IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-132
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX8201IRO1S_C
    8200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,888.27
    $4,888.27
    Each
       $4,888.27
    Supplier Number: QTX8201IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-134
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX3201IRO1S_C
    3200 g
    60 mg
    182×182 mm
    100 mg
    ≤0.9 s
    50 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $3,772.22
    $3,772.22
    Each
       $3,772.22
    Supplier Number: QTX3201IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-098
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX3202IMO1S_C
    3200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $6,004.31
    $6,004.31
    Each
       $6,004.31
    Supplier Number: QTX3202IMO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-100
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
  • QTX3202IRO1S_C
    3200 g
    6 mg
    182×182 mm
    10 mg
    ≤0.9 s
    5 mg
    215×377×95 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $4,888.27
    $4,888.27
    Each
       $4,888.27
    Supplier Number: QTX3202IRO1S
    Spécifications:
    More
    Specifications
    Cat. No.
    CA77507-102
    Operating temperature range
    10…30 °C
    Plug type
    Interchangeable country-specific plug-in AC adaptors
    Communication interface
    USB UTL;USB Device
    Voltage
    100 – 240 V
Accessories
Informations sur le produit
  • Catalog #
    Availability
    Price
  • Data cable, mini USB|RS232, 25-pin
    Data cable, mini USB|RS232, 25-pin
    CA76412-910
    Log in pour la disponibilité
    Each - $93.41