About this item
The Select DNA cloning Golden Gate Assembly BsmBI kits are compatible with the Type IIS restriction enzymes BsmBI. Please choose the appropriate kit based on the number of fragments being assembled.
- Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
- Supports user-provided linearized vector
- Length: 0.3 to 1.8 kb
- Supports up to >5 fragment assembly using BsmBI
- Compatible with BioXp® 3250
The Select DNA cloning Golden Gate Assembly kits are compatible with the Type IIS restriction enzyme BsmBI. The user will need to select the number of fragments they want to assemble, either 1-4 fragment assembly or 5 fragment assembly.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.