About this item
The Select DNA cloning and amplification kits generate a minimum of 10 µg of amplified DNA from fragments of 0.3 to 1.8 kb using the BioXp® 3250.
- Accepted input: Telesis Bio DNA fragments, IDT gBlocks™, Twist Biosciences fragments
- Supports user-provided linearized vector
- Length: 0.3 to 1.8 kb
- Yield: >10 µg/well
- Compatible with BioXp® 3250
The BioXp® Select DNA cloning and amplification kit contains Gibson Assembly® and other reagents necessary to synthesize at least 10 µg of amplified DNA from fragments and linearized vectors supplied by the user.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.