BioXp® Tip Kits
Supplier: TELESIS BIO
Jump to:
À propos de cet article
BioXp® system tips in various kit types and sizes.
- Free of human DNA
- Rnase/Dnase free
- PCR inhibitor free
- Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
Ordering information: Verify the tip requirements for the BioXp® kit that you are ordering.
16 Options Available Below
Product Details & Documents
BioXp® system tips in various kit types and sizes.
- Free of human DNA
- Rnase/Dnase free
- PCR inhibitor free
- Automated
Different kit types and sizes will require different quantities of 50 and 200 µl racks.
Ordering information: Verify the tip requirements for the BioXp® kit that you are ordering.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 12207KT
- Supplier Number: 12208KT
- Supplier Number: 12209KT
- Supplier Number: 12210KT
- Supplier Number: 12030KT
- Supplier Number: 12025KT
- Supplier Number: 12201KT
- Supplier Number: 12212KT
- Supplier Number: 12204KT
- Supplier Number: 12214KT
- Supplier Number: 12202KT
- Supplier Number: 12205KT
- Supplier Number: 12211KT
- Supplier Number: 12203KT
- Supplier Number: 12206KT
- Supplier Number: 12213KT