Laboratory Exhaust Canopies
Fournisseur: Eagle Group
Certificats
À propos de cet article
Stainless steel laboratory exhaust canopies are used to exhaust laboratory work spaces.
- 16 gauge type 304 stainless steel construction
- Fully welded seams
- Polished interior and exterior
- Steel rod suspension brackets
- Integral duct collar
- Full-perimeter condensation edge
These canopies are used for ventilation of heat, steam or non toxic materials with low vapor density and not for storage or mixing of hazardous chemicals. Type 304 stainless construction provides strength and durability. Available in island or wall mounted style.
Frequently Bought Together