Quick-Dry Foam Squeegees, Perfex | Avantor
Quick-Dry Foam Squeegees, Perfex
Supplier:   Perfex
Quick-Dry Foam Squeegees, Perfex
Supplier:  Perfex
Certificates
Quick-Dry Foam Squeegees, Perfex
Quick-Dry Foam Squeegees, Perfex
2 Options Available Below
Product Details & Documents
Twin-blade neoprene rubber squeegees are specifically designed for use on uneven and irregular surfaces
Note: Not recommended for abrasive surfaces.
Feature a durable polymer channel and flexible blades that quickly wipe surfaces dry. Lightweight, nonmarring, and easy to clean with warm water. Resistant to water, oil, grease, detergents, sanitizers, and solvents.
Ordering information: Polymer adjustable handle sold separately.
Product Family Options
Informations sur le produit
  • Désignation
    Longueur
    Availability
    Price
  • Foam Squeegee Blade
    61 cm (24")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $150.89
    $150.89
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $1,508.90
    $1,508.90
    Each
       $150.89
    Case of 10
       $1,508.90
    Supplier Number: 9724
  • Foam Squeegee Blade
    45.7 cm (18")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $125.41
    $125.41
    Log in pour la disponibilité
    Case of 10
    Case of 10 - $1,254.10
    $1,254.10
    Each
       $125.41
    Case of 10
       $1,254.10
    Supplier Number: 9718
Accessories
Informations sur le produit
  • Catalog #
    Handle Material
    Handle Color
    Availability
    Price
  • Adjustable Handle with Long Connector
    Adjustable Handle with Long Connector
    53200-100
    Polymer
    White
    Log in pour la disponibilité
    Each - $141.82