PIG® Absorbent Packaging Bag | Avantor
PIG® Absorbent Packaging Bag
Supplier:   New Pig
PIG® Absorbent Packaging Bag
Supplier:  New Pig
Certificates
PIG® Absorbent Packaging Bag
PIG® Absorbent Packaging Bag
5 Options Available Below
Product Details & Documents
Secure your shipment from escaping liquid by sealing your container in this packaging bag with absorbent sheet.
  • Sealable plastic bag with absorbent sheet traps and soaks up liquid escaping from container
  • Soft-but-heavyweight sheet adds light cushioning and scratch protection
  • Built-in mat flap lets you fold mat over the top for added barrier from seal point; tab also makes it easy to find the opening
  • Absorbs up to 1.41 ounces per bagged absorbent
  • Absorbent-bag combination works on most free liquids, including solvents, oils and water-based liquids
  • Durable multi-layer construction holds together, even when saturated
Product Family Options
Informations sur le produit
  • Désignation
    W×L
    Weight
    Color
    Packaging
    Availability
    Price
  • Absorbent packaging bag
    3×5"
    1.5 lbs.
    Grey
    50 bags per box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $116.86
    $116.86
    Box of 50
       $116.86
    Supplier Number: PKG017
  • Absorbent packaging bag
    4×6"
    1.8 lbs.
    Grey
    50 bags per box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $122.35
    $122.35
    Box of 50
       $122.35
    Supplier Number: PKG018
  • Absorbent packaging bag
    6×9"
    3.9 lbs.
    Grey
    50 bags per box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $153.37
    $153.37
    Box of 50
       $153.37
    Supplier Number: PKG019
  • Absorbent packaging bag
    10×12"
    8.5 lbs.
    Grey
    50 bags per box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $297.64
    $297.64
    Box of 50
       $297.64
    Supplier Number: PKG020
  • Absorbent packaging bag
    12×15"
    13.75 lbs.
    Grey
    50 bags per box
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 50
    Box of 50 - $345.10
    $345.10
    Box of 50
       $345.10
    Supplier Number: PKG021