Push Brooms, Perfex
Push Brooms, Perfex
 :   Perfex
Push Brooms, Perfex
 :  Perfex
Push Brooms, Perfex
Push Brooms, Perfex
Synthetic brushes are designed to exceed the rigorous demands of today’s controlled environments
Polypropylene fibers are fused onto a high impact-resistant polymer block to eliminate premature fiber fallout that could contaminate collection areas. Lite-N-Tite® handle/broom head connection system locks handle securely in place. Brush and handle are nonconductive and will not absorb bacteria, liquids, or odors. Both are unaffected by water, petroleum products, detergents, sanitizers, and solvents. Durable, lightweight, sanitary adjustable handle, made of high-quality polyester and composite materials, reduces strain and transfers maximum energy with minimum exertion.
Ordering information: Brush heads are available in two sizes and five colors for light/medium and heavy sweeping tasks. A special extra-heavy model is also available. Brush heads and handles sold separately.
Product Family Options
Soft Sweep
  • Désignation
    Bristle Material
    Bristle Color
    Head Width
  • Push Broom Head, Soft Sweep, 46L x 8W cm (18 x 3")
    PP
    Blue
    46 cm (18")
    Each
    Each - $66.36
    $66.36
    Each
       $66.36
     : 2618B
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-162
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3")
    PP
    Blue
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2624B
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-170
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 46L x 8W cm (18 x 3")
    PP
    Green
    46 cm (18")
    Each
    Each - $66.36
    $66.36
    Each
       $66.36
     : 2618G
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-288
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3")
    PP
    Green
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2624G
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-292
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 46L x 8W cm (18 x 3")
    PP
    Red
    46 cm (18")
    Each
    Each - $114.59
    $114.59
    Each
       $114.59
     : 2618R
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-160
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3")
    PP
    Red
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2624R
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-168
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 46L x 8W cm (18 x 3")
    PP
    Yellow
    46 cm (18")
    Each
    Each - $66.36
    $66.36
    Each
       $66.36
     : 2618Y
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-290
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3")
    PP
    Yellow
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2624Y
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-294
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 46L x 8W cm (18 x 3")
    PP
    White
    46 cm (18")
    Each
    Each - $66.36
    $66.36
    Each
       $66.36
     : 2618W
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-164
    Small Business Enterprise
  • Push Broom Head, Soft Sweep, 61L x 8W cm (24 x 3")
    PP
    White
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2624W
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-166
    Small Business Enterprise
Stiff Sweep
  • Désignation
    Bristle Material
    Bristle Color
    Head Width
  • Push Broom Head, Stiff Sweep, 46L x 8W cm (18 x 3")
    PP
    Blue
    46 cm (18")
    Each
    Each - $114.61
    $114.61
    Each
       $114.61
     : 2518B
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-174
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 61L x 8W cm (24 x 3")
    PP
    Blue
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2524B
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-184
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 46L x 8W cm (18 x 3")
    PP
    Green
    46 cm (18")
    Each
    Each - $68.00
    $68.00
    Each
       $68.00
     : 2518G
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-296
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 61L x 8W cm (24 x 3")
    PP
    Green
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2524G
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-300
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 46L x 8W cm (18 x 3")
    PP
    Yellow
    46 cm (18")
    Each
    Each - $66.36
    $66.36
    Each
       $66.36
     : 2518Y
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-298
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 61L x 8W cm (24 x 3")
    PP
    Yellow
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2524Y
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-302
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 46L x 8W cm (18 x 3")
    PP
    White
    46 cm (18")
    Each
    Each - $117.38
    $117.38
    Each
       $117.38
     : 2518W
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-176
    Small Business Enterprise
  • Push Broom Head, Stiff Sweep, 61L x 8W cm (24 x 3")
    PP
    White
    61 cm (24")
    Each
    Each - $75.06
    $75.06
    Each
       $75.06
     : 2524W
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-180
    Small Business Enterprise
Rough Sweep
  • Désignation
    Bristle Material
    Bristle Color
    Head Width
  • Push Broom Head, Rough Sweep, 46L x 10W cm (18 x 4")
    PP
    Orange
    46 cm (18")
    Each
    Each - $71.10
    $71.10
    Each
       $71.10
     : 2418
    Small Business Enterprise
    Specifications
    Cat. No.
    22940-188
    Small Business Enterprise
Accessories
  •  
    Description
    Handle Material
    Handle Color
  • 53200-100
    Adjustable Handle with Long Connector
    Polymer
    White
    Each - $141.82
  •  
    Description
    Handle Material
    Handle Color
  • 22940-104
    Fixed-Length Handle
    Polymer
    White
    Each - $77.26
  •  
    Description
    Dimensions
    Handle Length
  • 11310-906
    Hardwood Handle, 60", Natural
    2.5 cm (1") Diameter
    152 cm (60")
    Each - $19.76