Swivel Pad Holder and Scrubbing Pads, Perfex
: Perfex
Swivel pad holder is designed to twist and turn for cleaning hard-to-reach areas
Its velcro base holds pads securely in place. Attach a polymer adjustable handle to the pad holder to clean floors, baseboards, walls, windows, and ceilings. Also, ideal for cleaning fine surfaces, including sinks, vinyl, and stainless steel.
: USDA and FDA Approved materials.
: Pad Holder, pads, and handle are sold separately.
Frequently Bought Together