Multi-Purpose Brushes, Perfex
Supplier:   Perfex
Multi-Purpose Brushes, Perfex
Supplier:  Perfex
Certificates
Multi-Purpose Brushes, Perfex
Multi-Purpose Brushes, Perfex
5 Options Available Below
Product Details & Documents
These multi-purpose brushes contain moderately stiff polyester fiber.
  • Surround-set and flared fiber maintains surface contact at any angle and pprevents brush block from damaging surfaces
  • Durable fiber maintains shape and is visually coded to isolate areas of use
  • Impact-resistant polymer brush block withstands soaking and will not rot, decay, chip or crack
  • Non-conductive with superb abrasion and chemical resistance
  • Unaffected by water, grease, petroleum products, detergents, sanitizer, and solvents
Excellent for scrubbing irregular surfaces, as well as floors, walls, and ceilings.
 : Brush heads and handle sold separately.
Product Family Options
Informations sur le produit
  • Désignation
    Bristle Material
    Bristle Color
    Longueur totale
    Availability
    Price
  • Brush Head
    Polyester
    Blue
    10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $172.27
    $172.27
    Each
       $172.27
    Supplier Number: 3040B
  • Brush Head
    Polyester
    Green
    10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $170.33
    $170.33
    Each
       $170.33
    Supplier Number: 3040G
  • Brush Head
    Polyester
    Red
    10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $168.83
    $168.83
    Each
       $168.83
    Supplier Number: 3040R
  • Brush Head
    Polyester
    White
    10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $193.69
    $193.69
    Each
       $193.69
    Supplier Number: 3040W
  • Brush Head
    Polyester
    Yellow
    10"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $169.91
    $169.91
    Each
       $169.91
    Supplier Number: 3040Y
Accessories
Informations sur le produit
  • Catalog #
    Handle Material
    Handle Color
    Availability
    Price
  • Adjustable Handle with Short Connector
    Adjustable Handle with Short Connector
    53200-102
    Polymer
    White
    Log in pour la disponibilité
    Each - $149.14