AlphaSorb® HC Wipers
AlphaSorb® HC Wipers
Supplier:   Texwipe
AlphaSorb® HC Wipers
Supplier:  Texwipe
Certificates
AlphaSorb® HC Wipers
3 Options Available Below
Product Details & Documents
AlphaSorb® HC (high capacity) two-ply, knitted, 100% polyester wipers are engineered to combine high sorbency with low particle and extractable levels.
  • Autoclavable
The large surface area is especially suited for use in the pharmaceutical and biotechnology industries where large, highly sorptive wipers are required for applying and removing cleaning, sterilizing, and disinfecting solutions.
Certifications: Suitable for use in ISO Class 3 (FED-STD-209E Class 1/M1.5) environments.
Ordering information: Laundered in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom and packed in SolventSafe Bag-Within-A-Bag® packaging.
Specifications for Product Family
Specifications
  • Catalog No:
  • TW-TX2409
  • TWTX2412
  • TWTX2418
  • Basis weight:
  • 240 g/m²
  • 240 g/m²
  • 240 g/m²
  • Absorbency:
  • 680 ml/m²
  • 680 ml/m²
  • 680 ml/m²
  • Extractables:
  • 0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
  • 0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
  • 0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
  • Fibres (>100 µm):
  • 1.4×10³/m²
  • 1.4×10³/m²
  • 1.4×10³/m²
  • Particle generation (0,5 - 5 µm) - ASTM E2090 test:
  • 45×10⁶/m²
  • 45×10⁶/m²
  • 45×10⁶/m²
  • Particle generation (5 - 100 µm) - ASTM E2090 Test:
  • 9×10⁵/m²
  • 9×10⁵/m²
  • 9×10⁵/m²
  • Ions - chloride:
  • 0.09 ppm
  • 0.09 ppm
  • 0.09 ppm
  • Ions - potassium:
  • 0.25 ppm
  • 0.25 ppm
  • 0.25 ppm
  • Ions - sodium:
  • 0.1 ppm
  • 0.1 ppm
  • 0.1 ppm
Specifications
Product Family Options
Product Information
  • Packaging
    W×L
    Availability
    Price
  • 100 wipes/bag, 10 bags/case
    23×23 cm (9×9")
    Log in for availability
    Log in for availability
    Log in for availability
    Bag of 1,000
    Bag of 1,000 - $903.90
    $903.90
    New Customer Promotional Price
    Log in for availability
    Case of 1,000
    Case of 1,000 - $903.90
    $903.90
    New Customer Promotional Price
    Bag of 1,000
       $903.90
    Case of 1,000
       $903.90
    Supplier Number: TX2409
    Specifications:
    More
    Specifications
    Cat. No.
    TW-TX2409
    Basis weight
    240 g/m²
    Absorbency
    680 ml/m²
    Extractables
    0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
    Fibres (>100 µm)
    1.4×10³/m²
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    45×10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    9×10⁵/m²
    Ions - chloride
    0.09 ppm
    Ions - potassium
    0.25 ppm
    Ions - sodium
    0.1 ppm
  • 100 wipes/bag, 10 bags/case
    30.5×30.5 cm (12×12")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $186.40
    $186.40
    New Customer Promotional Price
    Log in for availability
    Case of 600
    Case of 600 - $1,118.40
    $1,118.40
    New Customer Promotional Price
    Pack of 100
       $186.40
    Case of 600
       $1,118.40
    Supplier Number: TX2412
    Specifications:
    More
    Specifications
    Cat. No.
    TWTX2412
    Basis weight
    240 g/m²
    Absorbency
    680 ml/m²
    Extractables
    0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
    Fibres (>100 µm)
    1.4×10³/m²
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    45×10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    9×10⁵/m²
    Ions - chloride
    0.09 ppm
    Ions - potassium
    0.25 ppm
    Ions - sodium
    0.1 ppm
  • 100 wipes/bag, 10 bags/case
    45.7×45.7 cm (18×18")
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $512.00
    $512.00
    New Customer Promotional Price
    Log in for availability
    Case of 300
    Case of 300 - $1,536.00
    $1,536.00
    New Customer Promotional Price
    Pack of 100
       $512.00
    Case of 300
       $1,536.00
    Supplier Number: TX2418
    Specifications:
    More
    Specifications
    Cat. No.
    TWTX2418
    Basis weight
    240 g/m²
    Absorbency
    680 ml/m²
    Extractables
    0.02 g/m² using deionized water, 0.10 g/m² using isopropyl alcohol
    Fibres (>100 µm)
    1.4×10³/m²
    Particle generation (0,5 - 5 µm) - ASTM E2090 test
    45×10⁶/m²
    Particle generation (5 - 100 µm) - ASTM E2090 Test
    9×10⁵/m²
    Ions - chloride
    0.09 ppm
    Ions - potassium
    0.25 ppm
    Ions - sodium
    0.1 ppm