ContactUsLinkComponent
ZymoBIOMICS Lysis Solution, Zymo Research
Catalog # 76358-806
Supplier:  Zymo Research
CAS Number:  
undefined
ZymoBIOMICS Lysis Solution, Zymo Research
Catalog # 76358-806
Supplier:  Zymo Research
Supplier Number:  D4300-1-150
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    ZymoBIOMICS Lysis Solution
  • Size:
    150 ml
  • Cat. no.:
    76358-806
Specifications
About this item
ZymoBIOMICS Lysis Solution is designed to be used with the ZymoBIOMICS DNA extraction kits. Used in combination with the ZR BashingBead Lysis Tubes, it assists with microbial lysis. It is certified to be low-bioburden to ensure minimal background contaminants for low abundance microbiome samples.