Storage and Assay Microplates | Avantor
Storage and Assay Microplates
Storage and Assay Microplates
Certificates
Storage and Assay Microplates
Storage and Assay Microplates
4 Options Available Below
Product Details & Documents
For biological assays, low storage temperatures, and filtrate collection applications.
  • Compatible with all major automation platforms and benchtop devices
  • No risk feasibility testing – test a sample in your application
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Molded with chemical resistant polymers, for maintained performance with prolonged exposure to aggressive solutions and solvents
  • Sealable with heat and adhesive seals, or cap mats, for superior performance
  • Low temperature tolerant for refrigerator or freezer storage
  • Autoclavable, can be sterilized at 121 °C, 15 psi for 20 minutes
  • Available in a large range of well volumes, from 58 µl to 10 ml for application scale-up or scale-down
  • Complementary fit to Agilent filter microplates for centrifugation stacking
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent supplies collection and storage assay microplates for a wide range of reaction chemistries, biological assays, low storage temperatures, and filtrate collection applications. These microplates are molded with medical-grade, chemical-resistant polymers and are available in 24, 48, 96, or 384-well configurations of variable volumes. Agilent also supplies mat seals and lids for these microplates.
Product Family Options
Informations sur le produit
  • Désignation
    Matière
    Volume
    Hauteur
    Maximum Well Volume
    Bottom Style
    Sterility
    Well Shape
    Availability
    Price
  • Storage/reaction microplate, 96-well
    PP
    0.5 ml
    14.3 mm
    528.36 µl
    Round
    Non sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $256.61
    $256.61
    Pack of 25
       $256.61
    La quantité minimale de commande est de 1
    Supplier Number: 204600-100
  • Storage/reaction microplate, 96-well
    PP
    1 ml
    32 mm
    1.28 ml
    Round
    Non sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $282.00
    $282.00
    Pack of 25
       $282.00
    La quantité minimale de commande est de 1
    Supplier Number: 203426-100
  • Storage/reaction microplate, 96-well, irradiated. Suggested seal/lid 202497-100
    PP
    1 ml
    32 mm
    1.28 ml
    Round
    Sterile
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $316.00
    $316.00
    Pack of 25
       $316.00
    La quantité minimale de commande est de 1
    Supplier Number: 204357-100
  • Storage/reaction microplate, 96-well. Suggested seal/lid 201158-100, 201160-100
    PP
    2 ml
    44 mm
    2.38 ml
    Pyramid
    Non sterile
    Square
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $291.00
    $291.00
    Pack of 25
       $291.00
    La quantité minimale de commande est de 1
    Supplier Number: 201240-100