Diamond® Essentials™ Graduated Measuring Cylinders
À propos de cet article
Graduated measuring cylinders offer the precise fluid measurements required in many laboratory operations. These cylinders are available in both Class A accuracy for precision work, and Class B accuracy for general laboratory work. Engineered with easy to read molded graduations, hexagonal bases, wide openings, and tapered pour spouts for simple and efficient use.
- Available in translucent polypropylene (PP) or clear polymethylpentene (PMP)
- Manufactured from virgin, USP class VI resins
- Chemically resistant to most acids, bases, and many common solvents
- Molded metric graduation scales per Class A or B accuracy standards
Working temperature range: 4 to 60 ºC. Calibrated to contain (TC) (In) at 20 ºC.
Certifications: Complies with FDA 21-CFR and European Pharmacopeia. Comply with DIN 12681 and ISO 6706 standards.
16 Options Available Below
Product Details & Documents
Graduated measuring cylinders offer the precise fluid measurements required in many laboratory operations. These cylinders are available in both Class A accuracy for precision work, and Class B accuracy for general laboratory work. Engineered with easy to read molded graduations, hexagonal bases, wide openings, and tapered pour spouts for simple and efficient use.
- Available in translucent polypropylene (PP) or clear polymethylpentene (PMP)
- Manufactured from virgin, USP class VI resins
- Chemically resistant to most acids, bases, and many common solvents
- Molded metric graduation scales per Class A or B accuracy standards
Working temperature range: 4 to 60 ºC. Calibrated to contain (TC) (In) at 20 ºC.
Certifications: Complies with FDA 21-CFR and European Pharmacopeia. Comply with DIN 12681 and ISO 6706 standards.
Product Family Options
Class A, Polymethylpentene (PMP)
Informations sur le produit
- AvailabilityPrice
- Supplier Number: 3705-100
- Supplier Number: 3705-250
- Supplier Number: 3705-500
- Supplier Number: 3705-1M
- Supplier Number: 3705-2M
Class B, Polymethylpentene (PMP)
Informations sur le produit
- AvailabilityPrice
- Supplier Number: 3703-10
- Supplier Number: 3703-25
- Supplier Number: 3703-50
- Supplier Number: 3703-250
- Supplier Number: 3703-500
- Supplier Number: 3703-1M
- Supplier Number: 3703-2M
Class B, Polypropylene (PP)