Securline® Surgical Skin Marker, Sterile, Aspen Surgical
52877-453
- Description:Surgical skin marker, fine/broad tip, violet
- Cat. no.:52877-453
- Supplier No.:1002-00-PDG
Marker features non-tattooing, surgical prep-resistant Gentian Violet ink that is nontoxic to the epidermic layer of the skin
Non-scratching. Bulk-packed.