PYREX® Serum Bottles, Corning | Avantor
PYREX® Serum Bottles, Corning
Supplier:   Corning
PYREX® Serum Bottles, Corning
Supplier:  Corning
Certificates
PYREX® Serum Bottles, Corning
PYREX® Serum Bottles, Corning
2 Options Available Below
Product Details & Documents
Designed for the handling and storage of sterile culture media and sera when the stability of glass is of prime importance.
  • Fits a #8 rubber stopper
  • Stopper not included
  • Bottle withstands repeated wet or dry autoclaving
These serum bottles are made of Pyrex® borosilicate glass.
Also suitable for storing distilled water and standard solutions. Necks are tooled for uniform rubber stopper fit and increased mechanical strength.
Product Family Options
Informations sur le produit
  • Capacité
    Availability
    Price
  • 4 L (1 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 8
    Case of 8 - $2,230.00
    $2,230.00
    Case of 8
       $2,230.00
    Supplier Number: 1585-4L
  • 9 L (2.4 gal.)
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4
    Case of 4 - $1,800.00
    $1,800.00
    Case of 4
       $1,800.00
    Supplier Number: 1585-9L