Low-Particle Bottles and Jugs, Thermo Scientific
Glass containers are certified to contain fewer than 5 particles per ml larger than 0.5 µm.
- Class 100/10 glass
- Certificate of Analysis is included with each shipment
- Available in clear or amber
- Individual bottles double bagged to maintain integrity until point of use
- 4 L jugs suitable for hazardous shipping in conjunction with combination packaging
- Recommended for low-particle applications
Ideal for preparation and storage of standards, off-line D.I. water and chemical sampling for particle analysis, and storage and packaging of chemicals, reagents, and powders. Amber glass protects light-sensitive chemicals. Containers are assembled with low-shedding, polypropylene caps with chemically-inert fluoropolymer-resin faced liners that do not contain adhesives. Exceeds USP specifications for particle content of containers.
: All containers are double-bagged in a ISO Class 5 (FED-STD-209E Class 100/M3.5) cleanroom. Supplied with Certificate of Analysis.
Frequently Bought Together