SP Wilmad-LabGlass Burettes with Precision Bore and Straight Stopcock with PTFE Plug, Class A, SP Industries
The top of these Class A burettes are funnel shaped to accommodate a rubber stopper and glass tubing to fill from a delivery tip via suction
Made of borosilicate glass with durable colored scale, both sizes have 2mm stopcock with PTFE plug.
Frequently Bought Together