SP Wilmad-LabGlass Precision Bore Class A Burettes with PTFE Stopcock, SP Industries
Fournisseur: SP Industries
Certificats
À propos de cet article
Made of borosilicate glass, these precision type, Class A burets have a straight 2mm PTFE stopcock
Calibration is in compliance with ASTM E 694 specifications.
Certifications: Reference No. ASTM E 287 Laboratory Glass Graduated Burets.
Frequently Bought Together