VWR® Alkali Burets, Automatic Self-Zeroing, Class A, Fluoropolymer Resin Stopcock
Fournisseur: VWR International
Certificats
À propos de cet article
Precision-grade borosilicate glass buret assembly and receiving bottle.
- Fluoropolymer resin stopcock eliminates contamination from lubricants
- Prefilled with activated alumina, ready to use
- Easy-to-read white enamel graduations
Both the U-shaped and straight drying tubes are prefilled with activated alumina.
Ordering information: Shipped complete with fluoropolymer resin stopcock, alumina filled drying tubes, and receiving bottle.
Frequently Bought Together