VWR® Amber Burets, Automatic Self-Zeroing, Class A, Glass Stopcock
Borosilicate glass burets for use with light-sensitive liquids.
- Amber glass for UV protection
Burets and bottles are treated to reduce UV light penetration, and meet Class A specifications for burets.
: Shipped complete with buret, bottle, rubber bulb, and stopcock.
Frequently Bought Together