VWR® Burets, Automatic Self-Zeroing, Class A, with Stopcock
VWR® Burets, Automatic Self-Zeroing, Class A, with Stopcock
VWR® Burets, Automatic Self-Zeroing, Class A, with Stopcock
VWR® Burets, Automatic Self-Zeroing, Class A, with Stopcock
Borosilicate glass buret assembly with precision-grade white scale.
  • Specially designed for accuracy and economy
Assembly seats securely into [ST] ground glass bottle top. Manufactured to Class A specifications for burets.
 : Shipped complete with bottle, rubber bulb, and stopcock.
Product Family Options
  • Capacité
    Intervalle de graduation
    Reservoir Capacity
    ST Joint No.
    Stopcock Type
  • 25 ml
    0.10 ml
    1000 ml
    24/20
    Glass
    Each
    Each - 3 376,55 $
    3 376,55 $
    Each
       3 376,55 $
  • 25 ml
    0.10 ml
    1000 ml
    24/20
    Fluoropolymer resin
    Each
    Each - 3 855,42 $
    3 855,42 $
    Each
       3 855,42 $
  • 10 ml
    0.05 ml
    1000 ml
    24/40
    Glass
    Each
    Each - 3 873,86 $
    3 873,86 $
    Each
       3 873,86 $
  • 10 ml
    0.05 ml
    1000 ml
    24/40
    Fluoropolymer resin
    Each
    Each - 4 078,87 $
    4 078,87 $
    Each
       4 078,87 $
  • 50 ml
    0.10 ml
    2000 ml
    29/42
    Glass
    Each
    Each - 4 516,89 $
    4 516,89 $
    Each
       4 516,89 $
  • 50 ml
    0.10 ml
    2000 ml
    29/42
    Fluoropolymer resin
    Each
    Each - 5 238,40 $
    5 238,40 $
    Each
       5 238,40 $