About this item
Top-Track™ system increases storage capacity up to 50% and maximizes the usable storage area in a given space.
- Chrome-plated units are ideal for dry environments with minimal exposure to corrosives
- Type 304 stainless steel units provide maximum protection in corrosive environments
Metroseal 3™ units are made of electroplated carbon steel with a unique corrosion-resistant epoxy, an economical alternative to stainless steel units, and are coated with antimicrobial product protection which inhibits the growth of bacteria, mold, mildew, and fungi that cause odors, stains, and product degradation. Stationary end and stationary intermediate unit kits for single systems include four 218.4 cm (86") high posts and the necessary hardware for connecting track sets to two stationary end units or one stationary intermediate unit. Stationary end and stationary intermediate unit kits for double deep systems include eight 218.4 cm (86") high posts, the necessary hardware for connecting two stationary end units in the double deep configuration, and the necessary hardware for connecting track sets to two stationary end units or one stationary intermediate unit. A stationary intermediate unit is required when track length exceeds 6.4 m (21'). The stationary end and stationary intermediate units must contain a minimum of four shelves. The width of the stationary end and stationary intermediate units should be equal to or greater than the width of the mobile units.
Mobile unit kits for single systems include four 188 cm (74") posts, four casters with bumpers, four caster channels, and four roller assemblies. Mobile unit kits for double deep systems also include the necessary hardware to connect two mobile units in the double deep configuration. The recommended maximum load rating for a mobile unit is 408.2 kg (900 lbs.). For the mobile units to function properly, the floor area should be level, smooth, and free from large cracks and raised obstacles.
Track sets include two segments of track. Only one track set is required between stationary units. The wall mount bracket kit includes one pair of brackets to attach a Top-Track™ system to a wall. The stop plate kit includes stops and fasteners for one end of the track set.
Note: Super Erecta® or Super Erecta® Super Adjustable™ shelves must be purchased separately.
The overhead track system guides mobile units between stationary units. An access aisle can be opened between any two units as needed. This open access to all units promotes cleanliness and permits users to walk or push utility carts in between units, simplifying the material handling task.