Burets, Automatic Self-Zeroing, Dual Rotaflo® Stopcocks, ProScience | Avantor
Burets, Automatic Self-Zeroing, Dual Rotaflo® Stopcocks, ProScience
Supplier:   Proscience
Burets, Automatic Self-Zeroing, Dual Rotaflo® Stopcocks, ProScience
Supplier:  Proscience
Certificates
Burets, Automatic Self-Zeroing, Dual Rotaflo® Stopcocks, ProScience
2 Options Available Below
Product Details & Documents
Burets feature two Rotaflo stopcocks that help eliminate titration errors and never require lubrication
The Rotaflo® design ensures accuracy and contamination-free performance. Buret 87003-564 accepts 12mm O.D. tubing for side and fill arms. Buret 87003-566 accepts 14mm O.D. tubing. Both burets feature a durable white enamel scale.
Product Family Options
Informations sur le produit
  • Capacité
    Longueur
    Intervalle de graduation
    Availability
    Price
  • 25 ml
    560 mm
    0.1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,499.34
    $1,499.34
    Each
       $1,499.34
    Supplier Number: GS17545-505
  • 50 ml
    760 mm
    0.1 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,455.06
    $1,455.06
    Each
       $1,455.06
    Supplier Number: GS17545-549