Modular Long Handle Tool Hook Kit | Avantor
Modular Long Handle Tool Hook Kit
Modular long handle tool hook kit, steel, epoxy coated, Grey
Catalog # 77269-208
Supplier:   Triton Products
Fournisseur #:  1733
Modular Long Handle Tool Hook Kit
Catalog # 77269-208
Supplier:  Triton Products
Modular Long Handle Tool Hook Kit
Modular Long Handle Tool Hook Kit
Modular long handle tool hook kit, steel, epoxy coated, Grey
Price
$94.30
Log in pour la disponibilité
Each
Product Details & Documents
Storability® accommodates hard to store items and goes well beyond the lightweight and traditional wire shelf storage currently available.
  • Ideal for rakes, brooms, shovels, post hole diggers and any large, heavy long handle tools
  • Tool keeper hooks lock and hang directly on the top track using a lock on connection and a tightening screw to assure the hooks stay put
  • Use additional 1733 tool holders to expand across a wall area
  • Each heavy rod holds up to 50 lbs.
  • Constructed of 14-gauge heavy-duty gray epoxy coated steel to ensure long life
  • Assembled size is 33W×17³/₄"H
Storability® modular wall storage is the ideal way to store and organize unlimited types of large and heavy items securely on a wall in the maintenance room, factory, shop or any area that requires heavy-duty and dependable storage solutions. Storability offers the broadest choice of accessories to accommodate hard to store items that goes well beyond lightweight and traditional wire shelf storage currently available. Storability offers many modular accessory styles that each mount to the Storability frame system to keep your hard to store items organized. Each accessory item is easy to install and provides more ways to store items than ever. All Storability accessory storage items lock on to the frame to keep them in place and will never fall off when your items are being removed or returned to their place. Each Storability modular wall accessory is easily removed and repositioned on the frame in seconds as requirements change. Customize your expandable system at any time.
Ordering information: Limited lifetime warranty.
Delivery information: Mounting hardware included.
Spécifications
  • Color:
    Grey
  • Désignation:
    Modular long handle tool hook kit, steel, epoxy coated