PYREX® Volumetric Flasks, Screw Cap, Class A, Corning®
   Options durables disponibles
Supplier:   Corning
PYREX® Volumetric Flasks, Screw Cap, Class A, Corning®
Supplier:  Corning
   Options durables disponibles
Certificates
PYREX® Volumetric Flasks, Screw Cap, Class A, Corning®
3 Options Available Below
Product Details & Documents
Made of PYREX® borosilicate glass.
Flasks have an expanded open end to facilitate transfer of crystals, salts and powders. Standard GPI thread screw cap has an inert, cone-shaped, polyethylene liner for leakproof closure. Conforms to Class A capacity tolerances.
Specifications for Product Family
Spécifications
  • Catalog No:
  • 89091-262
  • 89091-064
  • 89091-438
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
  • I-RECs, ISO 14001 - Monterrey
Spécifications
Product Family Options
Informations sur le produit
  • Capacité
    Taille du bouchon
    Tolerance
    Availability
    Price
  • 25 ml
    18-410
    ±0.03 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $860.00
    $860.00
    Case of 12
       $860.00
    Supplier Number: 5650-25
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-262
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 100 ml
    18-410
    ±0.08 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $961.00
    $961.00
    Case of 12
       $961.00
    Supplier Number: 5650-100
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-064
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
  • 50 ml
    18-410
    ±0.05 ml
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 12
    Case of 12 - $927.00
    $927.00
    Case of 12
       $927.00
    Supplier Number: 5650-50
    Spécifications:
    Product available on GSA Advantage®
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    89091-438
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    I-RECs, ISO 14001 - Monterrey
    Product available on GSA Advantage®
      Low Manufacturing Impact
Accessories
Informations sur le produit
  • Catalog #
    Cap Size
    Availability
    Price
  • Replacement Phenolic GPI Threaded Cap
    Replacement Phenolic GPI Threaded Cap
    29619-534
    18-410
    Log in pour la disponibilité
    Case of 12 - $47.90