To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:ECL Western Blotting Substrate
- Size:50 mL (1.6 oz.)
- Cat. no.:CAPI32109
- For Membrane Area:400 cm²
- No. of Reactions:5 Mini-Gel blots
- Supplier No.:32109
A value-priced, entry-level peroxidase substrate for enhanced chemiluminescence (ECL) that directly replaces costlier products without the need to re-optimize conditions.
- Less cost than other ECL Substrates
- Switch from other entry-level ECL substrates without the need for optimization or protocol changes
This substrate provides reliability and performance equivalent to other standard ECL substrates for detection of horseradish peroxidase (HRP) enzyme activity. The luminol and peroxide reagent-formulations are identical to other available substrate products, so one can switch to this economical substrate without needing to optimize probing conditions or incubation protocols.