Nalgene® PMP Hydrometer Jar, Thermo Scientific
500 ml, 50 mm (2")
24661-255
: 6230-0500
Nalgene® PMP Hydrometer Jar, Thermo Scientific
24661-255
Hydrometer jar with cylinder of crystal-clear polymethylpentene (PMP)
Broad plastic base provides good stability. Suitable for use as lactometer cylinder.
Superior chemical resistance and safety. Withstands continuous usage at up to 135°C. Autoclavable. Tube ungraduated and without identification. No overflow cup required for reading.
- Ø ext.:50 mm (2")
- Capacité:500 ml
- Cat. No.:24661-255
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact