KIMAX® Double Metric Scale Cylinders with Bumper, Class B, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Calibrated to deliver and printed with a double white enamel ceramic scale
Designed from ASTM Specification E1272, Style 1, Class B requirements.
Cylinders with 25 through 2000mL capacities are supplied with a SAFE-GARD® bumper. The 10mL size has an enlarged, funnel-shaped top for ease of filling.
Spécifications
- Catalog No:
- 89001-196
- 89001-194
- 89001-190
- 89000-758
- 89001-188
- Form:
- Short
- Short
- Short
- Short
- Short