KIMAX® Graduated Cylinders, Class B, To Deliver, Blue Scale, Kimble Chase
Fournisseur: DWK Life Sciences (KIMBLE)
Certificats
À propos de cet article
Printed with durable blue enamel ceramic single metric scale
Designed from ASTM Specification E1272, Style 1, Class B requirements.
Cylinders with 25 through 2000mL capacities are supplied with a SAFE-GARD® bumper. The 10mL size has an enlarged funnel-shaped top for ease of filling.
Spécifications
- Catalog No:
- 89001-156
- 89001-152
- 89001-154
- 89001-148
- 89001-150
- 89001-144
- 89001-146
- 89001-142
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short