KIMAX® Reverse Scale Graduated Cylinders, Class A, To Deliver, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
Certificates
About this item
Cylinders are made of sturdy borosilicate glass and feature reverse metric graduations in white
Cylinders 25mL and larger are supplied with SAFE-GARD® bumpers, which are sturdy split rings made of yellow low-density polyethylene.
Specifications
- Catalog No:
- 89001-684
- 89001-682
- 89001-694
- 89001-696
- 89001-690
- 89001-692
- 89001-686
- 89001-688
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall