Reservoir Microplates
Reservoir Microplates
Certificates
Reservoir Microplates
Reservoir Microplates
17 Options Available Below
Product Details & Documents
Polypropylene reservoir microplates for automated liquid handlers and handheld pipettors.
  • Large range of cavity volumes, from 2.6 to 300 ml with specialized bottom geometries to optimize access and minimize dead volumes
  • Low binding surfaces for minimal losses of proteins, DNA/RNA, and other biologicals
  • Molded from chemical-resistant polymers, for maintained performance following prolonged exposure to aggressive solutions and solvents
  • Produced to ANSI/SLAS standards, for compatibility with popular labware and devices
  • Compatible with all major platforms and benchtop devices, making these reservoirs automation friendly
  • Low temperature tolerant, suitable for heating or cooling during use
  • Autoclavable, can be sterilized at 121 °C, 15 psi for 20 minutes
  • Fully customizable to fit your requirements
  • Manufactured in a DNAse/RNase-free ISO 9001 regulated facility
Agilent polypropylene reservoir microplates are available for a wide range of automated liquid handlers and handheld pipettors. These reservoir microplates are molded using high-quality, chemical-resistant polymers to ANSI/SLAS standard footprints, with optimized base geometries to minimize dead volumes. They are available with 1 to 24 cavities per reservoir plate, container heights between 19 and 44 mm, and with clear or black lids.
These reservoirs are recommended for both method scale-up and scale-down applications as well as for creating design of experiments (DoE) arrays.
Product Family Options
Single cavity polypropylene
Informations sur le produit
  • Désignation
    Hauteur
    Matière
    Maximum Volume
    Color
    Sterility
    Packaging
    Availability
    Price
  • Reservoir, single cavity, 96 pyramids base geometry, 86 ml
    19 mm
    PP
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $235.16
    $235.16
    Pack of 25
       $235.16
    Supplier Number: 201254-100
  • Reservoir, single cavity, 96 pyramids base geometry, irradiated, 86 ml
    19 mm
    PP
    White
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $330.00
    $330.00
    Pack of 25
       $330.00
    Supplier Number: 204484-100
  • Reservoir, single cavity, 12 column base geometry, 170 ml
    30.6 mm
    PP
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $307.00
    $307.00
    Pack of 25
       $307.00
    Supplier Number: 200686-100
  • Reservoir, single cavity, 12 column base geometry, irradiated, 170 ml
    30.6 mm
    PP
    White
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $339.00
    $339.00
    Pack of 25
       $339.00
    Supplier Number: 204363-100
  • Reservoir, single cavity, 8 row base geometry, 290 ml
    44 mm
    PP
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $357.00
    $357.00
    Pack of 25
       $357.00
    Supplier Number: 201252-100
  • Reservoir, single cavity, 8 row base geometry, irradiated, 290 ml
    44 mm
    PP
    White
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $393.00
    $393.00
    Pack of 25
       $393.00
    Supplier Number: 204504-100
  • Reservoir, single cavity, 12 column base geometry, 290 ml
    44 mm
    PP
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $273.50
    $273.50
    Pack of 25
       $273.50
    Supplier Number: 201250-100
  • Reservoir, single cavity, 12 column base geometry, irradiated, 290 ml
    44 mm
    PP
    White
    Sterile
    Individually bagged
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $396.00
    $396.00
    Pack of 25
       $396.00
    Supplier Number: 204093-100
  • Reservoir, single cavity, 96 pyramids base geometry, 300 ml
    44 mm
    PP
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $280.20
    $280.20
    Pack of 25
       $280.20
    Supplier Number: 201244-100
  • Reservoir, single cavity, 96 pyramids base geometry, 300 ml
    44 mm
    PP
    Black
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $399.00
    $399.00
    Pack of 25
       $399.00
    Supplier Number: 201246-100
  • Reservoir, single cavity, 96 pyramids base geometry, irradiated, 300 ml
    44 mm
    PP
    White
    Sterile
    Individually bagged
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $303.53
    $303.53
    Pack of 25
       $303.53
    Supplier Number: 204017-100
6 column polypropylene
Informations sur le produit
  • Désignation
    Hauteur
    Matière
    Maximum Volume
    Color
    Sterility
    Packaging
    Availability
    Price
  • Reservoir, 6 column, 24 pyramid base geometries, 47 ml/column
    44 mm
    PP
    282 ml
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $273.50
    $273.50
    Pack of 25
       $273.50
    Supplier Number: 201284-100
  • Reservoir, 6 column, 24 pyramid base geometries, irradiated, 47 ml/column
    44 mm
    PP
    282 ml
    White
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $396.00
    $396.00
    Pack of 25
       $396.00
    Supplier Number: 204284-100
12 column polypropylene
Informations sur le produit
  • Désignation
    Hauteur
    Matière
    Maximum Volume
    Color
    Sterility
    Packaging
    Availability
    Price
  • Reservoir, 12 column, pyramid base geometries, 7 ml/column
    19 mm
    PP
    84 ml
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $235.16
    $235.16
    Pack of 25
       $235.16
    Supplier Number: 201280-100
  • Reservoir, 12 column, pyramid base geometries, irradiated, 7 ml/column
    19 mm
    PP
    84 ml
    White
    Sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $339.00
    $339.00
    Pack of 25
       $339.00
    Supplier Number: 204365-100
  • Reservoir, 12 column, pyramid base geometries, 21 ml/column
    44 mm
    PP
    252 ml
    White
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $273.50
    $273.50
    Pack of 25
       $273.50
    Supplier Number: 201256-100
  • Reservoir, 12 column, pyramid base geometries, irradiated, 21 ml/column
    44 mm
    PP
    252 ml
    White
    Sterile
    Individually bagged
    Log in pour la disponibilité
    Log in pour la disponibilité
    Pack of 25
    Pack of 25 - $488.00
    $488.00
    Pack of 25
       $488.00
    Supplier Number: 204095-100