Nalgene® Settleometer Kit, Thermo Scientific
Settleometer Kit, Jar: 13.3dia. x 19.1H cm (53/16 x 71/2")
Tube: 1.7dia. x 11.9H cm (11/16 x 411/16")
Tube: 1.7dia. x 11.9H cm (11/16 x 411/16")
Catalog # 66185-009
Supplier: Thermo Fisher Scientific
Fournisseur #: 1010-0507
Nalgene® Settleometer Kit, Thermo Scientific
Catalog # 66185-009
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Economical kit contains everything needed to perform settleometer and centrifuge tests for monitoring sludge quality during the activated sludge process in wastewater treatment
Plastic components are chemically resistant to normal sludge samples (pH 3–10).
Ordering information: Kit consists of 2L jar with easy-to-read white graduations, cover, polypropylene paddle, ten 15mL conical polycarbonate centrifuge tubes with molded-in graduations and friction-fit polypropylene closures, instruction booklet, and data sheet.
Spécifications
- Désignation:Settleometer Kit
- Dimensions:Jar: 13.3dia. x 19.1H cm (53/16 x 71/2")
Tube: 1.7dia. x 11.9H cm (11/16 x 411/16") - Cat. No.:66185-009
- Environmental certification:ISO14001 (Certificate Registration No. EMS 752603)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Informations sur le produit
- Catalog #AvailabilityPrice
- Replacement Jar and Cover for Settleometer KitReplacement Jar and Cover for Settleometer Kit25408-090Log in pour la disponibilitéEach - $318.40