Pierce QuantaBlu™ Fluorogenic Peroxidase Substrates, Thermo Scientific
Catalog # CAPI15169
Supplier: Thermo Scientific
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Description:Fluorogenic Peroxidase Substrate Kit
- Cat. no.:CAPI15169
- Supplier No.:15169
Specifications
About this item
The substrate generates a blue fluorescent product upon reaction with peroxidase and does not photobleach.
- More sensitive than TMB, OPD or ABTS substrates
- Large dynamic range (4 log peroxidase concentration range)
- Working solution stable for 24 hours
- Large stokes shift; excitation/emission maxima of 325/420; range of 315 to 345/370 to 460
- Does not photobleach
QuantaBlu™ substrate also allows for stopped, nonstopped, and kinetic assays to be performed with low-level detection sensitivity and high signal-to-noise ratios.
Delivery: Kit includes QuantaBlu™ substrate solution (250 mL), QuantaBlu™ peroxide solution (30 mL), QuantaBlu stop solution (275 mL).