HPLC Remote Wall Bracket Kits, Justrite® | Avantor
HPLC Remote Wall Bracket Kits, Justrite®
Supplier:   Justrite
HPLC Remote Wall Bracket Kits, Justrite®
Supplier:  Justrite
Certificates
HPLC Remote Wall Bracket Kits, Justrite®
HPLC Remote Wall Bracket Kits, Justrite®
2 Options Available Below
Product Details & Documents
Ideal for applications utilizing safety disposal cans with disconnects.
HPLC filters can often be attached directly to the vapor release fitting on disposal cans. However, when this option is unavailable, remote wall brackets allow filters to be placed in a convenient and remote location. Disconnect fitting is valved to provide a safe, fully closed system that prevents vapor release when changing filters. Fittings accommodate 9.5 mm (3/8") O.D. tubing.
 : Bracket kits consist of a female quick disconnect fitting, a type 304 stainless steel holder, and two self-tapping mounting screws. Tubing not included.
Product Family Options
Informations sur le produit
  • Désignation
    Width
    Hauteur
    Availability
    Price
  • Wall Bracket with Polypropylene Fitting
    10.2 cm (4")
    12.7 cm (5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $355.15
    $355.15
    Prix promotionnel pour les nouveaux clients
    Each
       $355.15
    Supplier Number: 28150
  • Wall Bracket with Stainless Steel Fitting
    10.2 cm (4")
    12.7 cm (5")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $1,977.28
    $1,977.28
    Prix promotionnel pour les nouveaux clients
    Each
       $1,977.28
    Supplier Number: 28151