Nalgene® Separatory Funnels, Polypropylene, Squibb, Pear-Shape, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Translucent, leakproof funnels feature excellent chemical resistance and do not require lubrication
Compatible with a wide range of lab reagents. Separation lines between immiscible liquids are easy to observe. Funnel and screw closure are autoclavable; stopcock assembly may be chemically disinfected.
Frequently Bought Together