VITLAB® Short-Form Graduated Cylinders, SAN, BrandTech
Made of glass-clear styrene acrylonitrile (SAN).
- Short form for stability
- Molded graduations for easy measurement
Short-form design increases stability to help prevent spills. Circular base.
- Catalog No:
- 83008-852
- 83008-848
- 83008-844
- 83008-840
- 83008-836
- 83008-856
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
Frequently Bought Together