VITLAB® Graduated Cylinders, Polypropylene, Class B, Blue Scale, BrandTech
Fournisseur: Brandtech
Certificats
À propos de cet article
Made of polypropylene with ring-molded graduations overprinted with blue screening to simplify measurements.
- Translucent polypropylene is chemically resistant
- Blue screening for high contrast with many liquids
These VITLAB® graduated cylinders are made of polypropylene for durability and chemical resistance. Cylinders are highly translucent with molded graduations, and blue-screened overprint for easy meniscus readings. Hexagonal base resists rolling.
Certifications: Class B per DIN 12681.
Spécifications
- Catalog No:
- 83008-900
- 83008-870
- 83008-920
- 83008-930
- 83008-890
- 83008-860
- 83008-910
- 83008-880
- Form:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short