PYREX® VISTA™ Graduated Cylinders, Class A, To Contain, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, class A.
- Calibrated to contain (TC) in accordance with ASTM E-1272
- Hexagonal base with graduations in blue enamel
- 10 ml cylinder has a funnel top
Specifications
- Catalog No:
- 89090-456
- 89090-452
- 89090-448
- 89090-444
- 89090-464
- 89090-460
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey