PYREX® Graduated Cylinder, To Contain, with PYREX® Stopper, Corning
Supplier: Corning
Certificates
About this item
Borosilicate glass with hollow glass tapered stopper.
- Calibrated to contain (TC, In)
- Hexagonal base
- Ring marks at major graduations, marks and inscriptions in durable white enamel
Specifications
- Catalog No:
- 89090-632
- 89090-886
- 24760-189
- 24760-167
- 89090-924
- 89090-830
- 89090-620
- 89090-800
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey