PYREX® Lifetime Red™ Single Metric Scale Graduated Cylinders, To Contain, Corning
Fournisseur: Corning
Certificats
À propos de cet article
Calibrated ‘To Contain’ the scale and figures are durable white enamel, which stand out clearly against the red color-band on these lifetime red cylinders.
The 10 ml size has a funnel top. The 100 ml size is made in accordance with ASTM E-133 and E-1272 and is used in ASTM tests D-86 and D-216.
Ordering information: Bumper guards are supplied with 100 ml through 2000 ml inclusive.
Spécifications
- Catalog No:
- 89090-634
- 89090-888
- 89090-926
- 89090-832
- 89090-624
- 89090-622
- 89090-802
- 24710-544
- 89090-636
- 24710-522
- Form:
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Tall
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey