Staticide® Antistatic Neutralizer/Cleaner, ACL
Supplier:   ACL Staticide
Staticide® Antistatic Neutralizer/Cleaner, ACL
Supplier:  ACL Staticide
Certificates
Staticide® Antistatic Neutralizer/Cleaner, ACL
Staticide® Antistatic Neutralizer/Cleaner, ACL
3 Options Available Below
Product Details & Documents
A highly concentrated, biodegradable, antistatic cleaner that leaves no film or streaks and will not degrade a production environment’s antistatic properties
  • Exhibits a low foam level and constant detergency
  • The formulation contains no phosphates or alkalis and is safe for septic and sewage systems
Static charge decay: 0.2 s
 : Meets MIL-B-81705B, NFPA 56A. Electrostatic decay time is measured per FED Test Method 4046.
Product Family Options
Informations sur le produit
  • Taille
    Packaging
    Availability
    Price
  • 3.8 L (1 gal.)
    Bottle
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 4 (1/Itm/Bx)
    Case of 4 (1/Itm/Bx) - $374.50
    $374.50
    Case of 4 (1/Itm/Bx)
       $374.50
    Supplier Number: 4020-1
  • 204 L (54 gal.)
    Drum
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (54US Gal)
    Each (54US Gal) - $5,322.66
    $5,322.66
    Each (54US Gal)
       $5,322.66
    Supplier Number: 4020-2
  • 18.9 L (5 gal.)
    Pail
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each (5US Gal)
    Each (5US Gal) - $415.98
    $415.98
    Each (5US Gal)
       $415.98
    Supplier Number: 4020-5