Staticide® Acrylic Static-Dissipative Floor Finish, ACL Staticide
Fournisseur: ACL Staticide
Certificats
À propos de cet article
Suitable for electronic production lines, computer rooms, and other high-tech static-sensitive environments
- Tough, easy-to-use, nonmigratory acrylic copolymer is high gloss, slip- and detergent-resistant
- Can be spray-buffed for easy maintenance
- Can be machine-buffed and may be cleaned without loss of gloss on finish
For best results in curing, Staticide® neutralizer/cleaner should be used before applying finish (see Staticide® Antistatic Neutralizer/Cleaner, ACL series).
Charge generation <100 V
Static charge decay: 10 s
Surface resistance: –108 to 109 ohms/cm2
Certifications: Meets or exceeds MIL-B-81705B, NFPA 56A standards.
Frequently Bought Together