VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels
VWR® Buchner Filter Funnels
These chemical-porcelain Buchner filter funnels feature a fixed perforated plate and glazing inside and out, except for the rim.
  • Highly resistant to chemicals
  • Autoclavable
  • Glazed inside and out, except for the rim
Documents
Product Family Options
  • Capacité
    Hauteur
    Fits Paper Diameter
  • 4500 ml
    350 mm
    240 mm
    Case of 1
    Case of 1 - $885.63
    $885.63
    Case of 1
       $885.63
  • 2 ml
    64 mm
    13 mm
    Each
    Each - $85.53
    $85.53
    Case of 12
    Case of 12 - $1,026.38
    $1,026.38
    Each
       $85.53
    Case of 12
       $1,026.38
  • 30 ml
    89 mm
    35–40 mm
    Each
    Each - $83.22
    $83.22
    Case of 12
    Case of 12 - $998.69
    $998.69
    Pallet of 2112
    Pallet of 2112 - $175,769.30
    $175,769.30
    Each
       $83.22
    Case of 12
       $998.69
    Pallet of 2112
       $175,769.30
  • 87 ml
    101 mm
    50–55 mm
    Each
    Each - $100.49
    $100.49
    Case of 6
    Case of 6 - $602.93
    $602.93
    Each
       $100.49
    Case of 6
       $602.93
  • 186 ml
    143 mm
    70 mm
    Each
    Each - $130.85
    $130.85
    Case of 6
    Case of 6 - $785.10
    $785.10
    Each
       $130.85
    Case of 6
       $785.10
  • 320 ml
    160 mm
    90 mm
    Each
    Each - $174.87
    $174.87
    Case of 4
    Case of 4 - $699.49
    $699.49
    Each
       $174.87
    Case of 4
       $699.49
  • 550 ml
    195 mm
    110 mm
    Each
    Each - $187.48
    $187.48
    Case of 4
    Case of 4 - $749.94
    $749.94
    Each
       $187.48
    Case of 4
       $749.94
  • 700 ml
    202 mm
    110–124 mm
    Each
    Each - $306.98
    $306.98
    Case of 2
    Case of 2 - $613.95
    $613.95
    Each
       $306.98
    Case of 2
       $613.95
  • 1060 ml
    221 mm
    142–150 mm
    Each
    Each - $151.95
    $151.95
    Case of 2
    Case of 2 - $303.90
    $303.90
    Each
       $151.95
    Case of 2
       $303.90
  • 1860 ml
    270 mm
    185 mm
    Each
    Each - $679.41
    $679.41
    Case of 2
    Case of 2 - $1,358.83
    $1,358.83
    Each
       $679.41
    Case of 2
       $1,358.83