Nalgene® Low-Particulate Bottles, High-Density Polyethylene, Narrow Mouth, with Screw Caps, Thermo Scientific
Fournisseur: Thermo Fisher Scientific
Certificats
À propos de cet article
Packaging/shipping bottles are ideal for microelectronic reagents and biotech/pharmaceutical applications.
- Leakproof to 15 psi
High-density polyethylene containers have an average particle level of less than 30 particles per ml at 0.3 µm and greater.
Ordering information: Complete with polypropylene screw caps. Certificate of quality included with each case.
Frequently Bought Together