SP Bel-Art Precisionware® Narrow Mouth Bottles, Autoclavable Polypropylene, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Ideal for pharmaceutical or similar packaging, polypropylene Boston round bottles are clear and molded from FDA compliant material.
- Polypropylene construction
- Ribbed cap exteriors
- Autoclavable, with caps detached, to 121 °C (250 °F)
Polypropylene caps have a valve-seal design for a leakproof fit; deep square threads help prevent closure back-off during shipping. Ribbed cap exteriors are designed for torque closure and easy opening. Inner bottle surfaces are smooth and rounded for easy cleaning and thorough emptying.
Frequently Bought Together