VWR® Sterile Sample Bags
À propos de cet article
These sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
- Pliant polyethylene sterile tubing
- Highly resistant
- Wire closure for an airtight seal
- Round wire bags have two wires for closure
Bags are made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals. They feature a wire closure for an airtight seal, and wide-sealed bottoms for added leakproof protection. To use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Note: Do not use with alcoholic substances or where temperature exceeds 100 °C (212 °F).
Note: Do not use with alcoholic substances or where temperature exceeds 100°C (212°F).
: Comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
: Bags 89085-560 and 76289-492 are packaged in a double pouch for cleanroom applications.
42 Options Available Below
Product Details & Documents
These sample bags are ideal for transportation and storage of solids, semisolids, and liquids for environmental and carcass sampling, biomedical and pharmaceutical research, quality assurance procedures, food industry applications, and clinical and veterinary medicine.
- Pliant polyethylene sterile tubing
- Highly resistant
- Wire closure for an airtight seal
- Round wire bags have two wires for closure
Bags are made of pliant, highly resistant virgin polyethylene sterile tubing with no side seals. They feature a wire closure for an airtight seal, and wide-sealed bottoms for added leakproof protection. To use, tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.
Note: Do not use with alcoholic substances or where temperature exceeds 100 °C (212 °F).
Note: Do not use with alcoholic substances or where temperature exceeds 100°C (212°F).
: Comply with EPA, FDA, USDA, and HACCP regulatory and industry requirements.
: Bags 89085-560 and 76289-492 are packaged in a double pouch for cleanroom applications.
Certificates
Recherche par numéro de lotProduct Family Options
Round Wire Bags
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 60 ml (2 oz.)2.5 mil.7.6×12.7 cm (3×5")Pack of 500$236.91Case of 1000$473.82
- 150 ml (5 oz.)2.5 mil.7.6×17.8 cm (3×7")Discontinued-
- 450 ml (15 oz.)2.5 mil.11.4×22.9 cm (4.5×9")Discontinued-
- 1500 ml (50 oz.)3 mil.14×38.1 cm (5.5×15")Discontinued-
- 650 ml (22 oz.)3 mil.14×22.9 cm (5.5×9")Discontinued-
- 900 ml (30 oz.)2.5 mil.11.4×30.5 cm (4.5×12")Discontinued-
- 1500 ml (50 oz.)3 mil.14×38.1 cm (5.5×15")Discontinued-
- 1650 ml (55 oz.)3 mil.17.8×30.5 cm (7×12")Discontinued-
Round Wire Bags with Safety Tabs
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 2545 ml (86 oz.)3 mil.17.8×38.1 cm (7×15")Pack of 250$476.39Case of 1000$1,905.56
- 150 ml (5 oz.)2.5 mil.7.6×17.8 cm (3×7")Pack of 500$205.81Case of 1000$411.62
- 900 ml (30 oz.)2.5 mil.11.4×30.5 cm (4.5×12")Discontinued-
- 450 ml (15 oz.)2.5 mil.11.4×22.9 cm (4.5×9")Pack of 500$262.76Case of 1000$525.52
- 650 ml (22 oz.)3 mil.14×22.9 cm (5.5×9")Pack of 500$311.39Case of 1000$622.78
- 650 ml (22 oz.)4 mil.14×22.9 cm (5.5×9")Pack of 500$1,051.29Case of 1000$2,102.58
- 900 ml (30 oz.)2.5 mil.11.4×30.5 cm (4.5×12")Pack of 500$305.94Case of 1000$611.88
- 1500 ml (50 oz.)4 mil.14×38.1 cm (5.5×15")Pack of 500$1,145.92Case of 1000$2,291.84
- 1650 ml (55 oz.)4 mil.17.8×30.5 cm (7×12")Pack of 250$765.61Case of 1000$3,062.44
- 1650 ml (55 oz.)3 mil.17.8×30.5 cm (7×12")Pack of 250$233.65Case of 1000$934.60
Round Wire Bags with Safety Tabs and White Marking Areas
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 650 ml (22 oz.)4 mil.14×22.9 cm (5.5×9")Pack of 500$702.35Case of 1000$1,404.70
- 60 ml (2 oz.)2.5 mil.7.6×12.7 cm (3×5")Pack of 500$416.52Case of 1000$833.04
- 650 ml (22 oz.)3 mil.14×22.9 cm (5.5×9")Log in pour la disponibilitéPack of 500Pack of 500 - $572.54$572.54Prix promotionnel pour les nouveaux clientsLog in pour la disponibilitéCase of 1000Case of 1000 - $1,145.08$1,145.08Prix promotionnel pour les nouveaux clientsPack of 500$572.54Case of 1000$1,145.08
- 150 ml (5 oz.)2.5 mil.7.6×17.8 cm (3×7")Pack of 500$227.13Case of 1000$454.26
- 450 ml (15 oz.)2.5 mil.11.4×22.9 cm (4.5×9")Pack of 500$284.38Case of 1000$568.76
- 800 ml (27 oz.)2.5 mil.11.4×30.5 cm (4.5×12")Pack of 500$493.87Case of 1000$987.74
- 650 ml (22 oz.)3 mil.14×22.9 cm (5.5×9")Pack of 500$367.58Case of 1000$735.16
- 1650 ml (55 oz.)3 mil.17.8×30.5 cm (7×12")Pack of 250$266.60Case of 1000$1,066.40
- 1650 ml (55 oz.)4 mil.17.8×30.5 cm (7×12")Pack of 250$1,081.42Case of 1000$4,325.68
Combination Wire Bags with Safety Tabs
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 1650 ml (55 oz.)4 mil.17.8×30.5 cm (7×12")Pack of 250$516.42Case of 1000$2,065.68
- 1500 ml (50 oz.)3 mil.14×38.1 cm (5.5×15")Pack of 500$394.48Case of 1000$788.96
- 450 ml (15 oz.)2.5 mil.11.4×22.9 cm (4.5×9")Pack of 500$776.08Case of 1000$1,552.16
- 650 ml (22 oz.)3 mil.14×22.9 cm (5.5×9")Pack of 500$524.90Case of 1000$1,049.80
- 2250 ml (75 oz.)4 mil.25.4×30.5 cm (10×12")Pack of 250$517.89Case of 1000$2,071.56
- 4000 ml (135 oz.)4 mil.25.4×38.1 cm (10×15")Pack of 250$553.72Case of 1000$2,214.88
Combination Wire Bags with Safety Tabs and White Marking Areas
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 130 ml (4 oz.)2.5 mil.7.6×17.8 cm (3×7")Pack of 500$299.86Case of 1000$599.72
- 450 ml (15 oz.)2.5 mil.11.4×22.9 cm (4.5×9")Pack of 500$305.99Case of 1000$611.98
- 450 ml (15 oz.)2.5 mil.11.4×22.9 cm (4.5×9")Discontinued-
- 650 ml (22 oz.)3 mil.14×22.9 cm (5.5×9")Pack of 500$1,136.41Case of 1000$2,272.82
- 2250 ml (75 oz.)4 mil.25.4×30.5 cm (10×12")Pack of 250$676.78Case of 1000$2,707.12
- 1650 ml (55 oz.)4 mil.17.8×30.5 cm (7×12")Pack of 250$1,131.53Case of 1000$4,526.12
- 4000 ml (135 oz.)4 mil.25.4×38.1 cm (10×15")Pack of 250$708.90Case of 1000$2,835.60
Jumbo Flat Wire Bags
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 6.9 L (230 oz.)4 mil.30.5×35.6 cm (12×14")Pack of 50$102.59Case of 250$512.95
- 10.5 L (355 oz.)4 mil.30.5×61 cm (12×24")Case of 250$1,126.82Each$1,126.82
- 15 L (510 oz.)4 mil.38.2×61 cm (15×24")Log in pour la disponibilitéCase of 250Case of 250 - $818.89$818.89Prix promotionnel pour les nouveaux clientsCase of 250$818.89Each$818.89
Jumbo Poultry Rinse Bag with Tear-Off Opening
Informations sur le produit
- CapacitéÉpaisseurW×LAvailabilityPrice
- 48 L (1600 oz.)2 mil.61×61 cm (24×24")Discontinued-