HARLECO® Bouin Fixation Fluid, Sigma-Aldrich®, MilliporeSigma
Fournisseur: MilliporeSigma
Certificats
À propos de cet article
HARLECO® Tissue Fixative. Excellent for preserving delicate cytoplasmic structures such as cilia, mitotic spindles and astral radiations
- Prepared from dyes certified by the Biological Stain Commission (BSC)
- For In-Vitro Diagnostic Use
- For Professional Use
Tissue Fixative. Good for nuclei and chromosomes. Useful in mitosis, ogenesis and spermatogenesis studies. Excellent for preserving delicate cytoplasmic structures such as cilia, mitotic spindles and astral radiations.
Frequently Bought Together