VWR® Casseroles, Porcelain
Fournisseur: VWR International
Certificats
À propos de cet article
Casseroles are constructed of chemical porcelain with pouring lip and flat handle.
- Chemical porcelain construction
- For use in high-temperature applications
- Features pouring lip and flat handle
- Glazed interior and exterior
Glazed inside and out, except for rim and top of handle. For use in temperatures up to 1150°C (2101°F).
Frequently Bought Together