SAFEGUARD® Specimen Bags, Minigrip
SAFEGUARD® Specimen Bags, Minigrip
SAFEGUARD® Specimen Bags, Minigrip
Supplier:  Minigrip
Certificates
SAFEGUARD® Specimen Bags, Minigrip
SAFEGUARD® Specimen Bags, Minigrip
3 Options Available Below
Product Details & Documents
Reclosable three-wall specimen transport bags feature distinct black print on an orange background, with biohazard logo and check boxes labeled Freeze, Refrigerate, and Room Temperature.
Bags meet NCCLS M29-T2 specimen handling guidelines and OSHA 1910.1030 regulation for blood-borne pathogens containment and biohazard graphics.
The contents are accessible through a unique TearZone® feature that allows the bag to be opened without perforations that may compromise the safety of the bag or the health of the technician. Outside pouch provides protection and transportation for patient information. Bag with blue color block is color-coded to designate freezing or refrigeration.
 : The 15.2×22.9 cm (6×9") and 20.3×25.4 cm (8×10") bags are packed in perforated dispenser bags; the 30.5×38.1 cm (12×15") bags are packed in inner bags without the dispenser bag. Contact your Avantor sales representative for custom prints and sizes.
Product Family Options
Informations sur le produit
  • Désignation
    Épaisseur
    W×L
    Availability
    Price
  • Specimen bag
    2 mil.
    152×229 mm (6×9")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $248.05
    $248.05
    Case of 1,000
       $248.05
    Supplier Number: MGTZ69
  • Specimen bag
    2 mil.
    203×254 mm (8×10")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 1,000
    Case of 1,000 - $315.55
    $315.55
    Case of 1,000
       $315.55
    Supplier Number: MGTZ810
  • Specimen bag
    2 mil.
    305×381 mm (12×15")
    Log in pour la disponibilité
    Log in pour la disponibilité
    Case of 500
    Case of 500 - $267.99
    $267.99
    Prix promotionnel pour les nouveaux clients
    Case of 500
       $267.99
    Supplier Number: MGTZ1215